Learn More
Hirschmann™ Individual Calibration Certificate (DAkkS)
Product Code. 10468691
Click to view available options
For Use With (Equipment):
Burettes (2 mL, 5 mL, 10 mL, 25 mL, 50 mL, 100 mL)
Graduated Pipettes (0.1 mL, 0.2 mL, 0.5 mL, 1 mL, 2 mL, 3 mL, 5 mL, 10 mL, 20 mL, 25 mL, 50 mL)
Volumetric Flasks (1 mL, 2 mL, 3 mL, 5 mL, 10 mL, 20 mL, 25 mL, 50 mL, 100 mL, 150 mL, 250 mL, 500 mL, 1000 mL, 2000 mL, 5000 mL, 10000 mL)
akku-drive 20 + 50 mL
ceramus and ceramus-classic (fixed volumes 1 to 50 mL)
ceramus and ceramus-classic (variable volumes 0.2 to 60 mL)
labopette (fixed volume 0.5 μL to 5 mL)
labopette (variable volume 0.5 μL to 5 mL), labopettor
labopette 12-channel (5 to 250 μL)
labopette 8-channel (0.5 to 250 μL)
opus 10, 20 + 50 mL
solarus 10 + 20 + 50 mL
Unit Size:
Each
Product Code. 10468691
Brand: Hirschmann™ 9980135
Individual DAkkS calibration certificate with a individual serial number ID on the instrument and the certificate, tested and documented in the Hirschmann™ DAkks calibration laboratory.
  • Individual DAkkS calibration certificate
  • With serial number
  • Markings with instrument ID
  • With the DAkkS documentation
Specifications
For Use With (Equipment) Burettes (2 mL, 5 mL, 10 mL, 25 mL, 50 mL, 100 mL)
Description Individual Calibration Certificate
Product Type DAkkS Certificate
For Use With (Application) Hirschmann volumetric glass-ware delivered ex works