missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Indian Hedgehog/Ihh Antibody (9L16), mFluor Violet 450 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-22020MFV450
This item is not returnable.
View return policy
Description
Indian Hedgehog/Ihh Monoclonal antibody specifically detects Indian Hedgehog/Ihh in Human samples. It is validated for Western Blot, ELISA
Specifications
| Indian Hedgehog/Ihh | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 9L16 | |
| Western Blot, ELISA | |
| BDA1Indian hedgehog homolog (Drosophila), HHG2, HHG-2, Indian hedgehog, Indian hedgehog (Drosophila) homolog, Indian hedgehog homolog, indian hedgehog protein | |
| Human recombinant Indian Hedgehog/Ihh (N-terminus) | |
| 0.1 mL | |
| Stem Cell Signaling Pathway | |
| 3549 | |
| Store at 4C in the dark. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction