missing translation for 'onlineSavingsMsg'
Learn More

INCB054329, MedChemExpress

Product Code. p-200098386
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494638 10 mg 10mg
16485476 100 mg 100mg
16467357 5 mg 5mg
16463176 25 mg 25mg
16420937 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16494638 Supplier Medchem Express Supplier No. HY112504

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress INCB054329 is a potent BET inhibitor.

INCB054329 is a potent BET inhibitor.

Specifications

Chemical Name or Material INCB054329
CAS 1628607-64-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H16N4O3
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (287.06 mM)
SMILES O=C1NC2=C3N1[C@@H](C4=NC=CC=C4)COC3=C(C5=C(C)ON=C5C)C=C2
Molecular Weight (g/mol) 348.36
Formula Weight 348.36
Percent Purity 98.19%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.