Learn More
Cytiva Immobiline™ DryPlate for IEF
Product Code. 11723339 Shop All Cytiva Products
Change view
Click to view available options
:
Medium range; pH 4-7
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
11723339 Medium range; pH 4-7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11723339 Supplier Cytiva Supplier No. 80112828
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Obtain reproducible IEF separations with dry, precast gels containing immobilized pH gradients
Immobiline DryPlate precast gels contain preformed, immobilized, non-drifting pH gradients. The gels are rehydrated to a thickness of 0.5mm in the reswelling cassette accessory before use.
  • Medium range covering 3 pH units
  • Handle mechanically stable, plastic-backed gels with confidence
  • Immobiline technology produces an absolutely stable pH gradient, allowing extremely high voltages and high resolution over narrow pH ranges, even for low mobility proteins that require long focusing times
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) IEF
Type Precast Gel
Hazard Statement
  • EUH210-Safety data sheet available on request.
Supplemental information
  • UNKNOWN ACUTE-90 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 90% of components with unknown hazards to the aquatic environment.