Learn More
Imidacloprid-d4 (imidazolidine-4,4,5,5-d4), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Imidacloprid D4 (imidazolidin-4,4,5,5 D4) |
| CAS | 1015855-75-0 |
| Molecular Formula | C9 2H4 H6 Cl N5 O2 |
| Quantity | 0.05 g |
| Synonym | 1 H-Imidazol-4,5-d2-2-amine, 1-[(6-chloro-3-pyridinyl)methyl]-4,5-dihydro-4,5-d2-N-nitro-, Imidacloprid D4 (imidazolidin-4,4,5,5 D4), Imidacloprid-d4 |
| InChI Formula | InChI=1 S/C9H10ClN5O2/c10-8-2-1-7(5-12-8)6-14-4-3-11-9(14)13-15(16)17/h1-2,5 H,3-4,6H2,(H,11,13)/i3D2,4D2 |
| SMILES | [2 H]C1([2 H])N\C(=N/[N+](=O)[O-])\N(Cc2ccc(Cl)nc2)C1([2 H])[2 H] |
| IUPAC Name | (NE)-N-[1-[(6-chloropyridin-3-yl)methyl]-4,4,5,5-tetradeuterioimidazolidin-2-ylidene]nitramide |
| Molecular Weight (g/mol) | 259.69 |
| Formula Weight | 259.0774 g/mol |
| Show More |