Learn More
Velp Scientifica™ Imhoff Cone
Shop All Velp Products
Click to view available options
Material:
Glass
Plastic
Unit Size:
Each
Description
Velp Scientifica™ Imhoff Cone for use with FCS and JLT Series Flocculators.
- Transparent for visability
- Available in glass or plastic
- Available in graduated
- For use with Flocculators
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| For Use With (Equipment) | FCS, JLT |
| Material | Glass |