missing translation for 'onlineSavingsMsg'
Learn More

Imeglimin hydrochloride, MedChemExpress

Product Code. 16468757
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468757 5 mg 5mg
16486158 10 mg 10mg
16462297 50 mg 50mg
16456546 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16468757 Supplier Medchem Express Supplier No. HY14771A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Imeglimin hydrochloride (EMD 387008) is an oral glucose-lowering agent. Imeglimin also reduces reactive oxygen species (ROS) production, increases mitochondrial DNA and improves mitochondrial function.

Imeglimin hydrochloride (EMD 387008) is an oral glucose-lowering agent. Imeglimin also reduces reactive oxygen species (ROS) production, increases mitochondrial DNA and improves mitochondrial function.

Specifications

Chemical Name or Material Imeglimin hydrochloride
Physical Form Solid
CAS 775351-61-6
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C6H14ClN5
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym EMD 387008 hydrochloride
Solubility Information H2O : ≥ 50 mg/mL (260.88 mM) ∣DMSO : 25 mg/mL (130.44 mM; Need ultrasonic)
SMILES [H]Cl.NC1=N[C@@H](C)N=C(N(C)C)N1
Molecular Weight (g/mol) 191.66
Formula Weight 191.66
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.