missing translation for 'onlineSavingsMsg'
Learn More

Imazapic, MedChemExpress

Product Code. p-200097554
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16422296

Brand: Medchem Express HYB1860

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Imazapic is a selective herbicide for both the preand post-emergent control of some annual and perennial grasses and some broadleaf weeds

Imazapic is a selective herbicide for both the preand post-emergent control of some annual and perennial grasses and some broadleaf weeds

Specifications

Chemical Name or Material Imazapic
CAS 104098-48-8
Color White
Purity Grade Notes Research
Molecular Formula C14H17N3O3
Quantity 500 mg
Solubility Information DMSO : ≥ 100 mg/mL (363.24 mM)
SMILES O=C(C1=CC(C)=CN=C1C2=NC(C(C)C)(C)C(N2)=O)O
Molecular Weight (g/mol) 275.3
Formula Weight 275.3
Percent Purity 99.86%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.