missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imazapic, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Imazapic is a selective herbicide for both the preand post-emergent control of some annual and perennial grasses and some broadleaf weeds
Specifications
Specifications
| Chemical Name or Material | Imazapic |
| CAS | 104098-48-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C14H17N3O3 |
| Quantity | 100 mg |
| Solubility Information | DMSO : ≥ 100 mg/mL (363.24 mM) |
| SMILES | O=C(C1=CC(C)=CN=C1C2=NC(C(C)C)(C)C(N2)=O)O |
| Molecular Weight (g/mol) | 275.3 |
| Formula Weight | 275.3 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction