Learn More
Invitrogen™ Image-iT™ Low Density Lipoprotein Uptake Kit, Bodipy FL
Product Code. 15729230
Change view
Click to view available options
Quantity:
200 μL kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15729230 200 μL kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15729230 Supplier Invitrogen™ Supplier No. I34359
The Image-iT Low Density Lipoprotein Uptake Kit - Bodipy FL includes Bodipy FL dye-labeled LDL, unlabeled LDL, heparin, BSA, and assay buffer.
The kit was designed to specifically detect binding and uptake of LDL through the LDL receptor internalization pathway, allowing maximum control and flexibility in the experimental design. It can be used with fluorescence microscopy, flow cytometry, cell sorting, high-throughput analysis, and high content analysis.
• Optimized LDL uptake kit with two built-in controls
• Bodipy FL is detectable with GFP/FITC filter set (absorption peak: ∼515 nm, emission peak: ∼520 nm)
• Bodipy FL is fixable and multiplexable with orange, red, deep red, violet, and blue emission dyes, such as RFP, mCherry, DAPI, Hoechst, HCS LipidTOX, and LysoTracker Deep Red
Two controls are included in the kit: unlabeled LDL as an optional pretreatment to occlude cell surface receptors in advance of probing with the labeled construct, and heparin which will chelate the LDL in the solution phase outside of the cells, preventing binding and uptake. When cells are labeled in the absence of these pretreatment controls, a fluorescent signal indicates specific LDL receptor-mediated endocytosis.
TRUSTED_SUSTAINABILITY
Specifications
Format Kit
Quantity 200 μL kit
Conjugate Bodipy FL
Content And Storage • 1 x 200 μL Bodipy FL-LDL, 1 mg/mL
• 1 x 200 μL LDL (unlabeled), 2.5 mg/mL
• 1 x 50 mL LDL assay buffer, 1 X
• 1 vial heparin, 5 mg/mL
• 1 x 150 mg BSA
Store in refrigerator (2°C to 8°C) and protect from light.
Label Type BODIPY Dyes
Shipping Condition Wet Ice
Form Solution
Protein Subtype LDL
Product Line Molecular Probes