missing translation for 'onlineSavingsMsg'
Learn More

IM-12, MedChemExpress

Product Code. p-200100589
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495068 10 mg 10mg
16465806 100 mg 100mg
16461307 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16495068 Supplier Medchem Express Supplier No. HY12292

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IM-12 is an inhibitor of GSK-3β, with an IC50 of 53 nM, and also enhances Wnt signalling.

IM-12 is an inhibitor of GSK-3β, with an IC50 of 53 nM, and also enhances Wnt signalling.

Specifications

Chemical Name or Material IM-12
CAS 1129669-05-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H20FN3O2
Quantity 10 mg
Solubility Information DMSO : ≥ 54.9 mg/mL (145.47 mM)
SMILES O=C(C(NCCC1=CC=C(F)C=C1)=C2C3=C(C)NC4=C3C=CC=C4)N(C)C2=O
Molecular Weight (g/mol) 377.41
Formula Weight 377.41
Percent Purity 98.3%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.