Learn More
IM-12, MedChemExpress
Product Code. 16495068
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16495068
Brand: Medchem Express HY12292
MedChemExpress IM-12 is an inhibitor of GSK-3β, with an IC50 of 53 nM, and also enhances Wnt signalling.
IM-12 is an inhibitor of GSK-3β, with an IC50 of 53 nM, and also enhances Wnt signalling.
Specifications
Chemical Name or Material IM-12
CAS 1129669-05-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H20FN3O2
Quantity 10 mg
Solubility Information DMSO : ≥ 54.9 mg/mL (145.47 mM)
SMILES O=C(C(NCCC1=CC=C(F)C=C1)=C2C3=C(C)NC4=C3C=CC=C4)N(C)C2=O
Molecular Weight (g/mol) 377.41
Formula Weight 377.41
Percent Purity 98.3%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease