Includes
Protocol booklet with detachable, quick reference card, necessary components for specified number of preps, including prefilters and DNase I
- Recovers total RNA due to removal of genomic DNA through on-column DNase I treatment
- Can be used on cultured cells, tissue, yeast and hard-to-lyse bacterial strains
- Prefilters reduce lysate viscosity for maximum yield and purity
- Results can be obtained with small amounts of sample
- Binding capacity: 100μg
- Sample size: 30mg (animal tissue), up to 5 × 106 cells (cell culture)
- Elution volume: 40 to 120μL
- RNA Purity (A260/A280): 1.8 to 2.2
- Total time of preparation: <30 min/6 preps
Quantitative reverse transcriptase PCR (QRT-PCR), Northern blot, and microarray experiments
Specifications
Specifications
| Description | illustra RNAspin Mini RNA Isolation Kit is designed for rapid extraction of high-quality RNA from a wide range of sample types. |
| For Use With (Application) | Used for the isolation of high-quality total RNA from a wide range of source material including hard-to-lyse bacterial strains. Purified RNA is suitable for use in sensitive downstream applications such as quantitative reverse transcriptase PCR (qRT-PCR), |
| Includes | Protocol booklet with detachable, quick-reference guide and necessary components, including prefilters and DNase I |
| No. of Reactions | 50 Preps |
| Product Type | illustra RNAspin Mini Kit |
| RNase | 1.8-2.2 |
| Sample Volume | <30 mg |
| Test Time | < 30 min/6 preps |
| Yield | Up to 100μg |
Safety and Handling
Signal Word
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H302+H312+H332-Harmful if swallowed, in contact with skin or if inhaled
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P501-Dispose of contents/container to .