Learn More
Cytiva Ready-To-Go™ GenomiPhi V3 DNA Amplification Kit
Product Code. 12967160 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
24
96
480
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
12967160 96 Each
12957160 24 Each
12977160 480 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12967160 Supplier Cytiva Supplier No. 25660196
Highly efficient and representative whole-genome amplification with 40 to 60μg yield from nanogram amounts of DNA sample. Cytiva Lifescience™ illustra™ Ready-To-Go™ GenomiPhi V3 DNA Amplification Kit is part of the Phi29 DNA polymerase family of products.
  • Representative amplification of the whole genome.
  • Preformulated, predispensed, single-dose, lyophilized cakes for rapid reaction set-up and greater reproducibility.
  • Quick and simple automation-friendly protocol; no thermal cycler required.
  • Shipping and storage at room temperature provides easy access and reduced carbon footprint.
  • Yields high-quality DNA (e.g. for high-throughput genotyping and DNA archival).
  • The reaction mixture is provided as predispensed, single-dose, lyophilized cakes in either strips of 8 tubes, or 96-well plates.
  • Amplification is highly uniform over the entire genome so that locus representation remains extremely close to the original DNA sample.
  • Amplification is carried out with very high fidelity due to Phi29 DNA polymerase proofreading activity.
  • The average product length is over 10kb.
  • Procedure is quick, simple and does not require a thermocycler.
  • A typical DNA yield of 12 to 20μg DNA can be achieved in less than two hours from only 10ng of genomic DNA input.
  • The starting material for GenomiPhi reactions can be purified DNA from any commercial kit or homebrew method, or a nonpurified cell lysate may be used.
  • Protocols for the amplification of DNA from blood lysates and dried blood spots are included in the manual.
  • GenomiPhi amplified DNA is suitable for various applications such as genotyping (SNP, STR, array CGH), cloning, sequencing, and DNA archival.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) For whole genome amplification by isothermal strand displacement amplification; typical downstream applications are genotyping (SNP, STR, array CGH) and DNA archiving.
Includes Ready-To-Go GenomiPhi V3 cakes, 1 × 96-well plate; 2x Denaturation buffer, 1 × 1.2 ml; Control DNA (Lambda), 500 ng; 12 Strips of 8 domed caps
No. of Reactions 96
Hazard Statement
  • EUH210-Safety data sheet available on request.
Supplemental information
  • UNKNOWN ACUTE-72.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 72.5% of components with unknown hazards to the aquatic environment.