Learn More
Cytiva PuReTaq Ready-To-Go™ PCR Beads
Shop All Cytiva Products
Click to view available options
Format:
0.2mL Tube with hinged cap
0.2mL Tubes/plate
0.5mL Tube
No. of Reactions:
96
100
480
Volume:
200μL
500μL
Dimensione della confezione:
100 reactions
480 reactions
Each
Descrizione
- puReTaq DNA polymerase and high-purity reagents provide robust performance with very low levels of contaminating DNA
- Manufacturing process locks in performance for excellent reproducibility with less risk of pipetting errors and contamination
- Long-term ambient-temperature stability simplifies shipping and removes any need for freezer storage
- Verified for real-time PCR
- Ready-To-Go means simply add template DNA and primers and cycle
Specifica
Specifica
| Content And Storage | Room temperature stable beads containing ∼ 2.5 units of recombinant PuReTaq DNA Polymerase, dATP, dCTP, dGTP, dTTP, stabilizers, BSA and reaction Buffer, When a bead is reconstituted to a 25μL final volume, the concentration of each dNTP |
| Format | 0.2mL Tubes/plate |
| Quantity | 1 Kit |
| For Use With (Application) | Standard PCR |
| No. of Reactions | 96 |
| Volume | 200μL |
Sicurezza e movimentazione
Identificativo prodotto
- PuReTaq
- EUH210-Safety data sheet available on request.
- UNKNOWN ACUTE-73.3 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 73.3% of components with unknown hazards to the aquatic environment.
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto