Learn More
IL-9 Receptor Alpha (CD129) Mouse anti-Human, BUV661, Clone: AH9R7, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 750610
This item is not returnable.
View return policy
Description
IL-9 Receptor Alpha (CD129) Monoclonal antibody specifically detects IL-9 Receptor Alpha (CD129) in Human samples. It is validated for Flow Cytometry
Specifications
| IL-9 Receptor Alpha (CD129) | |
| Monoclonal | |
| Brilliant Ultraviolet 661 | |
| CD129; Interleukin-9 receptor; IL-9R alpha chain; IL-9Rα | |
| Human IL9R Transfected Cell Line | |
| RUO | |
| Human | |
| IgG2b κ |
| Flow Cytometry | |
| AH9R7 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV661 under optimal conditions that minimize unconjugated dye and antibody. |