Learn More
IL-9 Receptor Alpha (CD129) Mouse anti-Human, BUV661, Clone: AH9R7, BD Biosciences™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 750610
Product Code. 17221925
  • ONLINE EXCLUSIVE PRICE
    665.00 EUR
    663.59 EUR / 50µg
    Save 1.41 EUR
    0.21% Off
Description
IL-9 Receptor Alpha (CD129) Monoclonal antibody specifically detects IL-9 Receptor Alpha (CD129) in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-9 Receptor Alpha (CD129)
Monoclonal
Brilliant Ultraviolet 661
CD129; Interleukin-9 receptor; IL-9R alpha chain; IL-9Rα
Human IL9R Transfected Cell Line
RUO
Human
IgG2b κ
Flow Cytometry
AH9R7
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV661 under optimal conditions that minimize unconjugated dye and antibody.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions