missing translation for 'onlineSavingsMsg'
Learn More

IL-6 Rabbit anti-Human, Clone: 2000000000, Proteintech

Product Code. 17629826 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17629826 20 μL 20µL
17619826 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17629826 Supplier Proteintech Supplier No. 810771RR20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Antibody

IL-6 antibody apecifically detetects IL-6 antigen in Human Samples. It is validated for Western Blot, ELISA.

Specifications

Antigen IL-6
Applications Western Blot, ELISA
Clone 2000000000
Concentration 1000 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Gene Accession No. BC015511
Gene Symbols IL6
Host Species Rabbit
Immunogen IL-6 fusion protein Ag10565
Purification Method Protein A purification
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 3569
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.