missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-3R alpha/CD123 Antibody (7G3) - Chimeric - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
267.00 EUR - 635.00 EUR
Specifications
| Antigen | IL-3R alpha/CD123 |
|---|---|
| Clone | 7G3 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18671675
|
Novus Biologicals
NBP3-20160-0.025mg |
0.025 mg |
267.00 EUR
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18660205
|
Novus Biologicals
NBP3-20160-0.2mg |
0.2 mg |
635.00 EUR
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-3R alpha/CD123 Monoclonal antibody specifically detects IL-3R alpha/CD123 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, NeutralizationSpecifications
| IL-3R alpha/CD123 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174 | |
| The original antibody was generated by intraperitoneally immunizing BALB/c mice with 5 X lo6 COS cells transfected with the IL-3R a-chain in combination with adjuvant peptides. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| 7G3 | |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Neutralization | |
| Unconjugated | |
| Rabbit | |
| B Cell Development and Differentiation Markers, Cytokine Research, Oncogenes, Phospho Specific, Protein Kinase | |
| PBS | |
| 3563 | |
| IgG κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title