missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-17/IL-17A Antibody (CL10252), Novus Biologicals™
Mouse Monoclonal Antibody
299.00 EUR - 625.00 EUR
Specifications
| Antigen | IL-17/IL-17A |
|---|---|
| Clone | CL10252 |
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18662109
|
Novus Biologicals
NBP3-07997-100ul |
100 μg |
625.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18691947
|
Novus Biologicals
NBP3-07997-25ul |
25 μg |
299.00 EUR
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-17/IL-17A Monoclonal antibody specifically detects IL-17/IL-17A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| IL-17/IL-17A | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A | |
| This antibody was developed against a synthetic peptide corresponding to amino acids: HRNEDPERYPSVIWEAK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| CL10252 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity | |
| PBS, pH 7.2, containing 40% glycerol | |
| 3605 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title