missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-15/IL-15R Complex Monoclonal Antibody (GRW15PLZ), Functional Grade, eBioscience™
GREENER_CHOICE

Product Code. 15228817
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15228817 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15228817 Supplier Invitrogen™ Supplier No. 16815682

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Description: The monoclonal antibody GRW15PLZ recognizes the mouse IL-15/IL-15 receptor complex. Interleukin-15 (IL-15) is a 14 kDa proinflammatory protein that is has been shown to play a role in the activation of neutrophils, dendritic cells, and macrophages, and is essential to the development and survival of NK cells and CD8 T-cells. IL-15 activates Jak/Stat pathways by signaling through a heterotrimeric receptor. Two of the subunits of this receptor, beta and gamma, are shared with the IL-2 receptor, while the alpha subunit is unique to IL-15. Despite the expression of IL-15 mRNA in many cell types, the secreted protein is rarely detectable in biological samples. Recent research suggests that IL-15 is retained inside the cell and is only secreted when in complex with its unique receptor, IL-15R alpha. The cytokine/receptor complex may stay bound to the cell surface, where it can be trans-presented to cells expressing the beta gamma subunits of the receptor, or may be secreted in a soluble form that lacks the transmembrane domain of the receptor. The soluble IL-15/IL-15R complex is produced by mouse dendritic cells, fibroblasts, and macrophages and demonstrates a 10-100-fold increase in agonistic activity over IL-15 alone. The monoclonal antibody GRW15PLZ reacts with and inhibits the bioactivity of IL-15 and the IL-15/IL-15R complex. Applications Reported: The monoclonal antibody GRW15PLZ reacts with and inhibits the bioactivity of IL-15 and the IL-15/IL-15R complex. A...

Interleukin-15 (IL-15) is a 14 kDa proinflammatory protein that has been shown to play a role in the activation of neutrophils, dendritic cells, and macrophages, and is essential to the development and survival of NK cells and CD8 T-cells. IL-15 activates Jak/Stat pathways by signaling through a heterotrimeric receptor. Two of the subunits of this receptor, beta and gamma, are shared with the IL-2 receptor, while the alpha subunit is unique to IL-15.Despite the expression of IL-15 mRNA in many cell types, the secreted protein is rarely detectable in biological samples. Recent research suggests that IL-15 is retained inside the cell and is only secreted in complex with its unique receptor, IL-15R alpha. This chaperoning is required from the ER to Golgi through to secretion. The cytokine/receptor complex may stay bound to the cell surface, where it can be trans-presented to cells expressing the beta/gamma subunits of the receptor, or may be secreted in a soluble form that lacks the transmembrane domain of the receptor. The soluble IL-15/IL-15R complex is produced by mouse dendritic cells, fibroblasts, and macrophages, and demonstrates a 10-100-fold increase in agonistic activity over IL-15 alone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-15/IL-15R Complex
Applications Functional Assay, Neutralization
Classification Monoclonal
Clone GRW15PLZ
Concentration 1 mg/mL
Conjugate Functional Grade
Formulation PBS with no preservative; pH 7.2
Gene Il15
Gene Accession No. P48346, Q60819
Gene Alias AA690181; AI503618; CD215; Il15; IL-15; IL-15 receptor subunit alpha; Il15ra; IL-15RA; IL-15R-alpha; ILN; Interleukin; interleukin 15; interleukin 15 receptor, alpha chain; interleukin-15; Interleukin-15 receptor subunit alpha; sIL-15 receptor subunit alpha; Soluble interleukin-15 receptor subunit alpha
Gene Symbols Il15, Il15ra
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16168, 16169
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.