Learn More
Invitrogen™ IL-1 alpha Monoclonal Antibody (ILA8-H12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ M420A
Description
M420A targets IL-1 alpha in ELISA applications and shows reactivity with Human samples. The M420A immunogen is recombinant human IL-1 alpha. M420A detects IL-1 alpha which has a predicted molecular weight of approximately 18 kDa. This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).
IL-1 alpha (Interleukin-1 alpha) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 alpha is coded by the IL1A gene and signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 beta. IL1A is located on the q arm on chromosome 2 at position 13. IL-1 alpha plays an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses. IL-1 alpha activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 alpha is a pleiotropic cytokine involved in various immune responses, inflammatory processes, and hematopoiesis. Further, IL-1 alpha is produced by monocytes and macrophages as a proprotein, which is proteolytically processed and released in response to cell injury and induces apoptosis. The IL1A gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. IL1 alpha regulates the activities of NF-kappaB and mitogen-activated protein kinases. IL-1 alpha can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO). High levels of IL-1 alpha are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis.
Specifications
| IL-1 alpha | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with no preservative | |
| P01583 | |
| Il1a | |
| Recombinant human IL-1 alpha. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA | |
| ILA8-H12 | |
| Unconjugated | |
| Il1a | |
| CTLA8; hematopoietin-1; IL 1 a; IL 1α; IL1; IL-1 alpha; IL17; IL1A; Il-1a; IL-1alpha; IL1-ALPHA; IL1F1; IL1α; ILN; Interleukin; interleukin 1 alpha; interleukin 1, alpha; interleukin 17; interleukin 1a; interleukin 1-alpha; interleukin 1-alpha (AA 1 - 152); Interleukin1 alpha; interleukin-1 alpha; interleukin-1 alpha precursor; Interleukin-17A; preinterleukin 1 alpha; pre-interleukin-1 alpha; pro-interleukin-1-alpha; RP23-160G19.8 | |
| Mouse | |
| Protein G | |
| RUO | |
| 3552 | |
| -20°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.