Learn More
Invitrogen™ IL-1 alpha Monoclonal Antibody (ALF-161), Functional Grade, eBioscience™

Armenian Hamster Monoclonal Antibody
349.00 EUR
Specifications
| Antigen | IL-1 alpha |
|---|---|
| Clone | ALF-161 |
| Concentration | 1 mg/mL |
| Content And Storage | 4°C |
| Applications | Functional Assay, Immunostaining, Neutralization |
Description
Description: The ALF-161 antibody reacts with mouse interleukin-1alpha (IL-1alpha). Mouse IL-1alpha, also called Lymphocyte Activating Factor (LAF), Endogenous Pyrogen (EP), Leukocyte Endogenous Mediator (LEM), and Mononuclear Cell Factor (MCF), is a 17 kDa factor produced by a wide variety of cells, including macrophages, dendritic cells, T and B cells. IL-1alpha is mostly cell-associated, with 23% amino acid homology with IL-1beta. The immune regulatory role of IL-1alpha is exerted on a wide range of cells including lymphocytes, epithelial cells and fibroblasts. In vivo, it induces hypotension, fever, and acute phase response. Applications Reported:The ALF-161 antibody has been reported for use in ELISA capture, ELISPOT capture, neutralization, and intracellular staining for flow cytometry. Applications Tested: The Functional Grade Purified ALF-161 antibody has been tested by LAL assay to verify low endotoxin levels, by ELISA, and in bioassay for neutralization of mouse IL-1 alpha bioactivity. The ALF-161 antibody at 0.031 μg/mL has been found to inhibit by 50% the biological effects of 4.0 ng/mL mouse IL-1 alpha in a D10 cell proliferation assay. Detailed information and protocols about cytokine bioassays and in vitro cytokine neutralization using antibodies can be found in the BestProtocols® section. The ALF-161 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of mouse interleukin-1 alpha (IL-1 alpha) in combination with the biot...
IL-1 alpha (Interleukin-1 alpha) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 alpha is coded by the IL1A gene and signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 beta. IL1A is located on the q arm on chromosome 2 at position 13. IL-1 alpha plays an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses. IL-1 alpha activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 alpha is a pleiotropic cytokine involved in various immune responses, inflammatory processes, and hematopoiesis. Further, IL-1 alpha is produced by monocytes and macrophages as a proprotein, which is proteolytically processed and released in response to cell injury and induces apoptosis. The IL1A gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. IL1 alpha regulates the activities of NF-kappaB and mitogen-activated protein kinases. IL-1 alpha can stimulate the expression of IL6 and cyclooxygenase-2 (PTGS2/COX-2), as well as enhance the production of nitric oxide (NO). High levels of IL-1 alpha are associated with several chronic inflammatory diseases including rheumatoid arthritis, psoriasis and multiple sclerosis.Specifications
| IL-1 alpha | |
| 1 mg/mL | |
| Functional Assay, Immunostaining, Neutralization | |
| Functional Grade | |
| Armenian Hamster | |
| RUO | |
| PBS with no preservative; pH 7.2 | |
| P01582 | |
| 16175 | |
| Primary | |
| Affinity chromatography |
| ALF-161 | |
| 4°C | |
| Monoclonal | |
| Liquid | |
| IgG | |
| Mouse | |
| Il1a | |
| CTLA8; hematopoietin-1; IL 1 a; IL 1α; IL1; IL-1 alpha; IL17; IL1A; Il-1a; IL-1alpha; IL1-ALPHA; IL1F1; IL1α; ILN; Interleukin; interleukin 1 alpha; interleukin 1, alpha; interleukin 17; interleukin 1a; interleukin 1-alpha; interleukin 1-alpha (AA 1 - 152); Interleukin1 alpha; interleukin-1 alpha; interleukin-1 alpha precursor; Interleukin-17A; preinterleukin 1 alpha; pre-interleukin-1 alpha; pro-interleukin-1-alpha; RP23-160G19.8 | |
| Il1a | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.