Learn More
DiscoverX IL-1α
Product Code. 17352479
Encompass
Click to view available options
:
2 μg
Product Code. 17352479
Brand: DiscoverX 921153
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
IL-1α is an agonist for IL1R1. Provided as 2μg of dry powder alongside reconstitution buffer.
Interleukin 1, alpha (IL-1α) is a pleiotropic cytokine involved in inflammation, immunity, and hematopoiesis. It is a pro-apoptotic protein. Stimulates thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation, and fibroblast growth factor activity. Has pro-inflammatory properties, also promoting fever and sepsis, and has been implicated in multiple diseases related to these symptoms. Interacts with TNF, another pro-inflammatory cytokine. Acts as an agonist for IL1R1. Provided as 2μg of dry powder alongside reconstitution buffer.
Specifications
Quantity 2 μg
Product Type Control Ligands