Learn More
IKA™ VX 1 Shaker Hand Attachment
Shop All IKA Products
Click to view available options
Holds:
250 mL
Unit Size:
Each
Description
- For vessels up to 250mL
Specifications
Specifications
| Holds | 250 mL |
| Product Type | One Hand Attachment |