missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IHH Polyclonal Antibody

Product Code. 16578381
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16578381 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16578381 Supplier Invitrogen™ Supplier No. PA593107

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: GCFPAGAQVR LESGARVALS AVRPGDRVLA MGEDGSPTFS DVLIFLDREP HRLRAFQVIE TQDPPRRLAL TPAHLLFTAD NHTEPAARFR ATFASHVQPG QYVLVAGVPG LQPARVAAVS THVALGAYAP LTKHGTLVVE DVVASCFAAV ADHHLAQLAF WPLRLFHSLA WGSWTPGEGV HWYPQLLYRL GRLLLEEGSF HPLGMSGAGS; Positive Samples: Human placenta, Mouse kidney, Mouse liver, Mouse ovary, Rat liver, Rat lung, Rat brain; Cellular Location: Cell membrane, Extracellular side, Lipid-anchor, extracellular space.

Ihh encodes a member of the hedgehog family of secreted signaling molecules. Hedgehog proteins are essential regulators of a variety of developmental processes including growth, patterning and morphogenesis. The encoded protein specifically plays a role in bone growth an differentiation. Mutations in this gene are the cause of brachydactyly type A1 which is characterized by shortening or malformation of the phalanges. Mutations in this gene are also the cause of acrocapitofemoral dysplasia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IHH
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.51 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Ihh
Gene Accession No. P97812, Q14623
Gene Alias BDA1; HHG2; HHG-2; Ihh; Indian hedgehog; Indian hedgehog homolog; indian hedgehog protein; Indian hedgehog protein C-product; Indian hedgehog protein N-product; indian hedgehog protein; LOW QUALITY PROTEIN: indian hedgehog protein; Indian hedgehog signaling molecule; Indian hedgehog-like protein
Gene Symbols Ihh
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 202-411 of human IHH (NP_0021722).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16147, 3549, 84399
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.