Learn More
IgY++ (IgG)(H+L) Rabbit anti-Chicken, HRP, Secondary Antibody, Proteintech
Product Code. 17660307 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
1 mL
200 μL
Unit Size:
1mL
200µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17660307 200 μL 200µL
17650307 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17660307 Supplier Proteintech Supplier No. SA000016200UL
Rabbit anti-Chicken Affinipure Secondary Antibody
Specifications
Antigen IgY++ (IgG)(H+L)
Applications ELISA, Western Blot
Conjugate HRP
Formulation 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
Host Species Rabbit
Purification Method The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Quantity 200 μL
Primary or Secondary Secondary
Target Species Chicken
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid