Learn More
IgM, μ Chain Specific Goat anti-Mouse, HRP, Secondary Antibody, Proteintech
Product Code. 17690387 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
1 mL
200 μL
Unit Size:
1mL
200µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17690387 200 μL 200µL
17680387 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17690387 Supplier Proteintech Supplier No. SA000126200UL
Goat anti-Mouse Affinipure Secondary Antibody
Specifications
Antigen IgM, μ Chain Specific
Applications Western Blot
Conjugate HRP
Formulation 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
Host Species Goat
Purification Method The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Quantity 200 μL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid