missing translation for 'onlineSavingsMsg'
Learn More

IgG(H+L) Goat anti-Rabbit, AP, Secondary Antibody, Proteintech

Product Code. 17630317 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17630317 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17630317 Supplier Proteintech Supplier No. SA0000221ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat anti-Rabbit Affinipure Secondary Antibody

Specifications

Antigen IgG(H+L)
Applications ELISA, Western Blot
Conjugate Alkaline Phosphatase (AP)
Formulation 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
Host Species Goat
Purification Method The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Quantity 1 mL
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.