Learn More
IgG(H+L) Goat anti-Human, HRP, Secondary Antibody, Proteintech
Product Code. 17680297 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17680297 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17680297 Supplier Proteintech Supplier No. SA00001171ML
Goat anti-Human Affinipure Secondary Antibody
Specifications
Antigen IgG(H+L)
Applications ELISA, Western Blot
Conjugate HRP
Formulation 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
Host Species Goat
Purification Method The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Quantity 1 mL
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid