Specifications
| Antigen | IgG(H+L) |
| Applications | Immunofluorescence, Flow Cytometry |
| Conjugate | FITC |
| Formulation | 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA |
| Host Species | Donkey |
| Purification Method | The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. |
| Quantity | 200 μL |
| Primary or Secondary | Secondary |
| Target Species | Goat |
| Content And Storage | Store at -20°C. Stable for one year after shipment. |
| Show More |