Learn More
IgG Fraction Monoclonal Mouse Anti-Rabbit IgG, Light Chain Specific Mouse anti-Rabbit, HRP, Secondary Antibody, Proteintech
Product Code. 17670307 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17670307 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17670307 Supplier Proteintech Supplier No. SA000017L1ML
Mouse Monoclonal Antibody
IgG Fraction Monoclonal Mouse Anti-Rabbit IgG, Light Chain Specific Monoclonal antibody apecifically detetects IgG Fraction Monoclonal Mouse Anti-Rabbit IgG, Light Chain Specific antigen in Rabbit samples. It is validated for ELSIA, Western Blot.
Specifications
Antigen IgG Fraction Monoclonal Mouse Anti-Rabbit IgG, Light Chain Specific
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate HRP
Formulation 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
Host Species Mouse
Purification Method The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Quantity 1 mL
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid