Learn More
Epredia™ IgG Antibody
Designed for immunofluorescence staining, Epredia™ IgG, FITC Labeled Antibody allows for rapid and inexpensive assessment of tissues.
Brand: Epredia™ SH-1921-R2
Description
Host Species: Sheep
Species Reactivity: Reacts with human γ-chain IgG.
Immunogen: IgG from human serum
Positive Control: Normal tonsil fixed and processed in the same manner as the test specimen
Cellular Localization: Large aggregates found in tonsil, lymph node, Peyer's patch and spleen
Recommended for:
- Immunofluorescence
Note:
USA: RUO; Int'l: RUO
Specifications
| IgG | |
| Polyclonal | |
| FITC | |
| IgG from human serum | |
| IVD | |
| Primary | |
| Prediluted |
| Immunofluorescence | |
| Polyclonal | |
| Sheep | |
| 2 mL | |
| Cancer and Tumor Biology | |
| Human |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.