missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG, FcγSubclass 2c Specific Goat anti-Mouse, HRP, Secondary Antibody, Proteintech
Shop All Proteintech ProductsSpecifications
Specifications
| Antigen | IgG, FcγSubclass 2c Specific |
| Applications | Western Blot |
| Conjugate | HRP |
| Formulation | 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA |
| Host Species | Goat |
| Purification Method | The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. |
| Quantity | 200 μL |
| Primary or Secondary | Secondary |
| Target Species | Mouse |
| Content And Storage | Store at -20°C. Stable for one year after shipment. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?