Learn More
IgG, Fcγ Fragment Specific Goat anti-Mouse, HRP, Secondary Antibody, Proteintech
Product Code. 17650287 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
1 mL
200 μL
Unit Size:
1mL
200µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17650287 1 mL 1mL
17660287 200 μL 200µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17650287 Supplier Proteintech Supplier No. SA000011A1ML
Goat anti-Mouse Affinipure Secondary Antibody
Specifications
Antigen IgG, Fcγ Fragment Specific
Applications ELISA, Western Blot
Conjugate HRP
Formulation 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
Host Species Goat
Purification Method The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Quantity 1 mL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid