missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IGFL4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16674065
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16674065 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16674065 Supplier Invitrogen™ Supplier No. PA599821

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total IGFL4.

IGFL4 gene ontology (GO) annotation includes signaling receptor binding and extracellular space.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IGFL4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IGFL4
Gene Accession No. Q6B9Z1
Gene Alias IGF; IGF like family member 4; IGFL4; IGF-like family member 4; Insulin growth factor-like family member 4; Insulin like growth factor
Gene Symbols IGFL4
Host Species Rabbit
Immunogen A synthesized peptide derived from human IGFL4(Accession Q6B9Z1), corresponding to amino acid residues R89-I124.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 444882
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.