Learn More
IFNAR1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13083-1-AP-20UL
This item is not returnable.
View return policy
Description
Interferons are widely used therapeutic agents because of their anti tumor and antiviral effects and because of their modulatory effects on the immune system. These cytokines produce their effects by binding to the Type 1 Interferon and Receptor (IFNAR1). Down regulation of this receptor plays a key role in determining the magnitude and duration of cytokine signaling. This down regulation is thought to be influenced by phosphorylation of Serine 535 and 539 in the IFNAR1.Specifications
| IFNAR1 | |
| Polyclonal | |
| Unconjugated | |
| IFNAR1 | |
| AVP, CRF2 1, IFN alpha REC, IFN alpha/beta receptor 1, IFN R 1, IFNAR, IFNAR1, IFNBR, IFRC, Type I interferon receptor 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3454 | |
| -20°C | |
| Liquid |
| Western Blot, Flow Cytometry | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P17181 | |
| IFNAR1 | |
| IFNAR1 Fusion Protein Ag3739 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |