Learn More
Brady™ IDXPERT™ Vinyl Labels
Click to view available options
Color:
White, Red
Unit Size:
1 roll
Specifications
Specifications
| Certifications/Compliance | 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC |
| Description | IDXPERT Labels |
| Adhesive Type | Acrylic, Permanent, Cold-temperature |
| Coating | Powder |
| Color | White, Red |
| Finish | Gloss |
| Material | Vinyl |
| Product Type | IDXPERT Label |
| For Use With (Application) | Indoor and Outdoor Grade Facility and Safety Identification, Laboratory Identification |
| For Use With (Equipment) | iDXPERT, LabXpert Printer |
| Show More |