missing translation for 'onlineSavingsMsg'
Learn More

IDO-IN-4, MedChemExpress

Product Code. 16483637
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483637 50 mg 50mg
16460306 2 mg 2mg
16427628 10 mg 10mg
16420098 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16483637 Supplier Medchem Express Supplier No. HY18769

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress IDO-IN-4 is an indoleamine 2,3-dioxygenase 1 (IDO-1) inhibitor, extracted from patent WO2014150677A1, Compound example 1 enantiomer 1.

IDO-IN-4 is an indoleamine 2,3-dioxygenase 1 (IDO-1) inhibitor, extracted from patent WO2014150677A1, Compound example 1 enantiomer 1.

Specifications

Chemical Name or Material IDO-IN-4
Physical Form Solid
CAS 1629125-65-0
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C26H35N3O3
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (228.53 mM)
SMILES O=C(NC1=CC=C(C)C=C1)NC2=CC([C@@H](C3)[C@@H]3C(O)=O)=CC=C2N(CC(C)C)CC(C)C
Molecular Weight (g/mol) 437.57
Formula Weight 437.57
Percent Purity 98.11%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.