Learn More
iCRT 14, MedChemExpress
Product Code. 16499647
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499647 5 mg 5mg
16487108 10 mg 10mg
16473147 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16499647 Supplier Medchem Express Supplier No. HY16665
MedChemExpress iCRT 14 is a novel potent inhibitor of β-catenin-responsive transcription (CRT), with IC50 of 40.3 nM against Wnt responsive STF16 luciferase.
iCRT 14 is a novel potent inhibitor of β-catenin-responsive transcription (CRT), with IC50 of 40.3 nM against Wnt responsive STF16 luciferase.
Specifications
Chemical Name or Material iCRT 14
CAS 677331-12-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H17N3O2S
Quantity 5 mg
Solubility Information DMSO : ≥ 29 mg/mL (77.24 mM)
SMILES O=C(S/1)N(C2=CC=CC=C2)C(C1=C\C3=C(C)N(C(C)=C3)C4=CN=CC=C4)=O
Molecular Weight (g/mol) 375.44
Formula Weight 375.44
Percent Purity 98.73%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death