missing translation for 'onlineSavingsMsg'
Learn More
Learn More
iCRT 14, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
iCRT 14 is a novel potent inhibitor of β-catenin-responsive transcription (CRT), with IC50 of 40.3 nM against Wnt responsive STF16 luciferase.
Specifications
Specifications
| Chemical Name or Material | iCRT 14 |
| CAS | 677331-12-3 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C21H17N3O2S |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 29 mg/mL (77.24 mM) |
| SMILES | O=C(S/1)N(C2=CC=CC=C2)C(C1=C\C3=C(C)N(C(C)=C3)C4=CN=CC=C4)=O |
| Molecular Weight (g/mol) | 375.44 |
| Formula Weight | 375.44 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction