missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICH/USP Parenteral Combined-2 Target Elements: Au @ 100; Ir, Os, Pd, Pt, Rh, Ru @ 10 μg/mL in 15% HCl, VHG™
Description
ICH/USP Parenteral Combined-2 Target Elements: Au @ 100; Ir, Os, Pd, Pt, Rh, Ru @ 10 μg/mL in 15% HCl
Specifications
Specifications
| Chemical Name or Material | Iridium/Osmium/Palladium/Platinum/Rhodium/Ruthenium/Gold |
| CAS | 7439-88-5, 4-2-7440, 5-3-7440, 6-4-7440, 7440-16-6, 7440-18-8 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Iridium/Osmium/Palladium/Platinum/Rhodium/Ruthenium/Gold |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | Dilute HCl |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Iridium 10 μg/mL/Osmium 10 μg/mL/Palladium 10 μg/mL/Platinum 10 μg/mL/Rhodium 10 μg/mL/Ruthenium 10 μg/mL/Gold 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?