missing translation for 'onlineSavingsMsg'
Learn More

ICA, MedChemExpress

Product Code. p-200101634
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16491909

Brand: Medchem Express HY22044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ICA (N-(pyridin-2-yl)-4-(pyridin-2-yl)thiazol-2-amine) is a SK channel inhibitor that has antileishmanial activity with an IC50 of 2.1 μM.

ICA (N-(pyridin-2-yl)-4-(pyridin-2-yl)thiazol-2-amine) is a SK channel inhibitor that has antileishmanial activity with an IC50 of 2.1 μM.

Specifications

Chemical Name or Material ICA
Physical Form Solid
CAS 3374-88-7
Color Brown
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C13H10N4S
For Use With (Application) COVID-19-immunoregulation
Synonym N-[4-(2-Pyridinyl)-2-thiazolyl]-2-pyridinamine
Solubility Information DMSO : ≥ 49.5 mg/mL (194.64 mM)
SMILES C1(NC2=NC(C3=NC=CC=C3)=CS2)=NC=CC=C1
Molecular Weight (g/mol) 254.31
Formula Weight 254.31
Percent Purity 99.63%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.