Learn More
IC-87114, MedChemExpress
Product Code. 16485987
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16485987
Brand: Medchem Express HY10110
MedChemExpress IC-87114 is a potent and selective PI3Kδ inhibitor with IC50 of 0.5 μM.
IC-87114 is a potent and selective PI3Kδ inhibitor with IC50 of 0.5 μM.
Specifications
Chemical Name or Material IC-87114
CAS 371242-69-2
Color Gray
Purity Grade Notes Research
Molecular Formula C22H19N7O
Quantity 5 mg
Solubility Information DMSO : 10 mg/mL (25.16 mM; Need ultrasonic)
SMILES O=C1N(C(CN2C3=C(C(N)=NC=N3)N=C2)=NC4=CC=CC(C)=C14)C5=C(C)C=CC=C5
Molecular Weight (g/mol) 397.43
Formula Weight 397.43
Percent Purity 98.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease