missing translation for 'onlineSavingsMsg'
Learn More

Ibuprofen Sodium Salt Dihydrate, TRC

Product Code. 30137914
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137914 5 g 5g
30132548 50 g 50g
17922385 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137914 Supplier Toronto Research Chemicals Supplier No. TRCI4001555G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 527688-20-6
Molecular Formula C13 H17 O2 . 2 H2 O . Na
Molecular Weight (g/mol) 264.293
Synonym Benzeneacetic acid, α-methyl-4-(2-methylpropyl)-, sodium salt, hydrate (1:1:2) (ACI), Benzeneacetic acid, α-methyl-4-(2-methylpropyl)-, sodium salt, dihydrate (9CI), Ibuprofen sodium dihydrate, Sodium ibuprofen dihydrate
IUPAC Name sodium;2-[4-(2-methylpropyl)phenyl]propanoate;dihydrate
SMILES O.O.[Na+].CC(C)Cc1ccc(cc1)C(C)C(=O)[O-]

Specifications

Quantity 5 g
InChI Formula InChI=1S/C13H18O2.Na.2H2O/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15;;;/h4-7,9-10H,8H2,1-3H3,(H,14,15);;2*1H2/q;+1;;/p-1
Formula Weight 264.134
Percent Purity >95%
Chemical Name or Material Ibuprofen Sodium Salt Dihydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.