missing translation for 'onlineSavingsMsg'
Learn More

IBT6A, MedChemExpress

Product Code. p-200099717
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481677 50 mg 50mg
16468217 5 mg 5mg
16426086 100 mg 100mg
16425508 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16481677 Supplier Medchem Express Supplier No. HY13036A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress IBT6A is an impurity of Ibrutinib. IBT6A can be used in synthesis of IBT6A Ibrutinib dimer and IBT6A adduct. Ibrutinib is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM.

IBT6A is an impurity of Ibrutinib. IBT6A can be used in synthesis of IBT6A Ibrutinib dimer and IBT6A adduct. Ibrutinib is a selective, irreversible Btk inhibitor with an IC50 of 0.5 nM.

Spezifikation

Chemical Name or Material IBT6A
CAS 1022150-12-4
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C22H22N6O
Quantity 50 mg
Solubility Information DMSO : 50 mg/mL (129.38 mM; ultrasonic and warming and heat to 60°C)
SMILES NC1=C2C(N([C@H]3CNCCC3)N=C2C4=CC=C(OC5=CC=CC=C5)C=C4)=NC=N1
Molecular Weight (g/mol) 386.45
Formula Weight 386.45
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Mehr anzeigen Weniger anzeigen

Research purposes only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.