Learn More
Invitrogen™ iBright™ SAE Software for 21 CFR Part 11, single license
Product Code. 17150031 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 license
Unit Size:
Each
Product Code. 17150031
Brand: Invitrogen™ A49208
For use with the iBright Imaging System and supports compliance with 21 CFR Part 11 FDA guidelines for security, auditing, and electronic (SAE) signatures using the SAE console.
  • Authentication of user login information
  • Ability to set password guidelines and expiry dates
  • Lockout access and re-authentication requirements after a period of inactivity
  • System lockout upon unauthorized attempts to access software
  • Ability of administrator to define user roles and permissions
  • Verification of each user's permissions to perform predefined activities
  • Identification of data tampering
  • Generation of audit trail log
  • Tracking of changes and committed actions to user roles, account management, and system settings
  • Ability of administrator to determine functions required to input reason for commit
  • Administrator determination of e-signature requirements
  • Ability to review records and sign electronically
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) iBright Imaging System
Product Line iBright
Type Software License
Quantity 1 license
Software Type Software License