missing translation for 'onlineSavingsMsg'
Learn More

Ibiglustat (L-Malic acid), MedChemExpress

Product Code. p-200097186
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16463197

Brand: Medchem Express HY16743A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Ibiglustat (Venglustat) L-Malic acid is an orally active, brain-penetrant glucosylceramide synthase (GCS) inhibitor. Ibiglustat L-Malic acid can be used for the research of Gaucher disease type 3, Parkinson's disease associated with GBA mutations, Fabry disease, GM2 gangliosidosis, and autosomal dominant polycystic kidney disease.

Ibiglustat (Venglustat) L-Malic acid is an orally active, brain-penetrant glucosylceramide synthase (GCS) inhibitor. Ibiglustat L-Malic acid can be used for the research of Gaucher disease type 3, Parkinson's disease associated with GBA mutations, Fabry disease, GM2 gangliosidosis, and autosomal dominant polycystic kidney disease.

Specifications

Chemical Name or Material Ibiglustat (L-Malic acid)
CAS 1629063-78-0
Color White
Purity Grade Notes Research
Molecular Formula C24H30FN3O7S
Quantity 50 mg
Synonym Venglustat (L-Malic acid) SAR402671 (L-Malic acid) GZ402671 (L-Malic acid)
Solubility Information DMSO : ≥ 100 mg/mL (191.00 mM)
SMILES O=C(O[C@@H]1CN2CCC1CC2)NC(C)(C3=CSC(C4=CC=C(F)C=C4)=N3)C.OC(C[C@H](O)C(O)=O)=O
Molecular Weight (g/mol) 523.57
Formula Weight 523.57
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.