missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibiglustat (L-Malic acid), MedChemExpress
Description
Ibiglustat (Venglustat) L-Malic acid is an orally active, brain-penetrant glucosylceramide synthase (GCS) inhibitor. Ibiglustat L-Malic acid can be used for the research of Gaucher disease type 3, Parkinson's disease associated with GBA mutations, Fabry disease, GM2 gangliosidosis, and autosomal dominant polycystic kidney disease.
Specifications
Specifications
| Chemical Name or Material | Ibiglustat (L-Malic acid) |
| CAS | 1629063-78-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H30FN3O7S |
| Quantity | 5 mg |
| Synonym | Venglustat (L-Malic acid) SAR402671 (L-Malic acid) GZ402671 (L-Malic acid) |
| Solubility Information | DMSO : ≥ 100 mg/mL (191.00 mM) |
| SMILES | O=C(O[C@@H]1CN2CCC1CC2)NC(C)(C3=CSC(C4=CC=C(F)C=C4)=N3)C.OC(C[C@H](O)C(O)=O)=O |
| Molecular Weight (g/mol) | 523.57 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?