missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibandronate Sodium Salt, TRC
Specifications
Specifications
| CAS | 138844-81-2 |
| Molecular Formula | C9 H22 N O7 P2 . Na |
| Quantity | 10 mg |
| Synonym | Phosphonic acid, P,P'-[1-hydroxy-3-(methylpentylamino)propylidene]bis-, sodium salt (1:1) (ACI), Phosphonic acid, [1-hydroxy-3-(methylpentylamino)propylidene]bis-, monosodium salt (9CI), BM 21.0955, BM 21.0955Na, Bondronat, Bondronate, Boniva, Bonviva, Ibandronate sodium, P,P'-[1-Hydroxy-3-(methylpentylamino)propylidene]bisphosphonic acid monosodium salt, BM 21.0955, BM 21.0955Na, Bondronat, Bondronate, Boniva, Bonviva, Ibandronate Sodium, [1-Hydroxy-3-(methylpentylamino)propylidene]bisphosphonic Acid Sodium Salt; |
| InChI Formula | InChI=1S/C9H23NO7P2.Na/c1-3-4-5-7-10(2)8-6-9(11,18(12,13)14)19(15,16)17;/h11H,3-8H2,1-2H3,(H2,12,13,14)(H2,15,16,17);/q;+1/p-1 |
| SMILES | [Na+].CCCCCN(C)CCC(O)(P(=O)(O)O)P(=O)(O)[O-] |
| IUPAC Name | sodium;hydroxy-[1-hydroxy-3-[methyl(pentyl)amino]-1-phosphono-propyl]phosphinate |
| Molecular Weight (g/mol) | 341.211 |
| Formula Weight | 341.0769 |
| Chemical Name or Material | Ibandronate Sodium Salt |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?