Learn More
Thermo Scientific Chemicals Ibandronate sodium salt monohydrate
Product Code. 16378297 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 16378297
Brand: Thermo Scientific Chemicals 463150050
Request Bulk or Custom Format
CAS: 138926-19-9 | C9H24NNaO8P2 | 359.23 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 138926-19-9
Molecular Formula C9H24NNaO8P2
Molecular Weight (g/mol) 359.23
InChI Key VBDRTGFACFYFCT-UHFFFAOYNA-M
IUPAC Name sodium hydrate hydrogen {1-hydroxy-3-[methyl(pentyl)amino]-1-phosphonopropyl}phosphonate
SMILES O.[Na+].CCCCCN(C)CCC(O)(P(O)(O)=O)P(O)([O-])=O
Specifications
Melting Point 175.0°C to 177.0°C
Quantity 5 g
Formula Weight 359.23
Chemical Name or Material Ibandronate sodium salt monohydrate